ContactUsLinkComponent
SFTS virus Gn Protein, His Tag (GNN-S52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BA47-1). 1 * 50 µG
Codice catalogo ACRBGNN-S52H3-50UG
Nome del fornitore:  ACROBIOSYSTEMS
SFTS virus Gn Protein, His Tag (GNN-S52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BA47-1). 1 * 50 µG
Codice catalogo ACRBGNN-S52H3-50UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together