Nipah virus Glycoprotein, His Tag (GLN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 71 - Thr 602 (Accession # Q9IH62-1). 1 * 500 µG
Codice catalogo ACRBGLN-N52H3500UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together