SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Hb) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion # QHD43416.1). The mutations C136F, Y144del, R190S, D215G, LA242-243del, Y449H, E484K, N501Y, D614G, H655Y, N67 1 * 500 µG
Codice catalogo ACRBSPN-C52HB500UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together