ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ha) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion # QHD43416.1). The mutations T95I, Y144S, Y145N, R346K, E484K, N501Y, D614G, P681H, D950N were identified in th 1 * 50 µG
Codice catalogo ACRBSPN-C52HA-50UG
Nome del fornitore:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ha) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion # QHD43416.1). The mutations T95I, Y144S, Y145N, R346K, E484K, N501Y, D614G, P681H, D950N were identified in th 1 * 50 µG
Codice catalogo ACRBSPN-C52HA-50UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together