Biotinylated Mouse CD40 Ligand, His,Avitag (CDL-M82H5) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548-1). 1 * 200 µG
Codice catalogo ACRBCDL-M82H5200UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together