ContactUsLinkComponent
Cynomolgus PADI2, His Tag (PA2-C5545) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Pro 665 (Accession # A0A2K5 TST4-1). 1 * 50 µG
  ACRBPA2-C5545-50UG
Cynomolgus PADI2, His Tag (PA2-C5545) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Pro 665 (Accession # A0A2K5 TST4-1). 1 * 50 µG
  ACRBPA2-C5545-50UG
Frequently Bought Together