ContactUsLinkComponent
Human SLITRK6, Mouse IgG2a Fc Tag (SL6-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Ser 608 (Accession # Q9H5Y7-1). 1 * 1 mg
Codice catalogo ACRBSL6-H5255-1MG
Nome del fornitore:  ACRO Biosystems
Human SLITRK6, Mouse IgG2a Fc Tag (SL6-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Ser 608 (Accession # Q9H5Y7-1). 1 * 1 mg
Codice catalogo ACRBSL6-H5255-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together