Biotinylated Human Siglec-7, His,Avitag (SG7-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Acc ession # Q9Y286-1). 1 * 200 µG
Codice catalogo ACRBSG7-H82E7200UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together