ContactUsLinkComponent
FITC-Labeled Human IL-15 R alpha, Fc Tag (ILA-HF255) is expressed from human 293 cells (HEK293). It contains AA Ile 31-Thr 205 (Acce ssion # Q13261-1). 1 * 25 µG
Codice catalogo ACRBILA-HF255-25UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human IL-15 R alpha, Fc Tag (ILA-HF255) is expressed from human 293 cells (HEK293). It contains AA Ile 31-Thr 205 (Acce ssion # Q13261-1). 1 * 25 µG
Codice catalogo ACRBILA-HF255-25UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together