ContactUsLinkComponent
Biotinylated Mouse IL-17C, His,Avitag (ILC-M82E9) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 194 (Acces sion # Q8K4C5-1). 1 * 200 µG
Codice catalogo ACRBILC-M82E9200UG
Nome del fornitore:  ACRO Biosystems
Biotinylated Mouse IL-17C, His,Avitag (ILC-M82E9) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 194 (Acces sion # Q8K4C5-1). 1 * 200 µG
Codice catalogo ACRBILC-M82E9200UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together