ContactUsLinkComponent
Biotinylated Mouse Midkine, His,Avitag (MIE-M82Q8) is expressed from human 293 cells (HEK293). It contains AA Lys 23 - Asp140 (Acces sion # P12025-1). 1 * 25 µG
Katalognr. ACRBMIE-M82Q8-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse Midkine, His,Avitag (MIE-M82Q8) is expressed from human 293 cells (HEK293). It contains AA Lys 23 - Asp140 (Acces sion # P12025-1). 1 * 25 µG
Katalognr. ACRBMIE-M82Q8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together