Biotinylated Human IL-2 R alpha, Fc,Avitag (ILA-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 ( Accession # P01589-1). 1 * 200 µG
Codice catalogo ACRBILA-H82F9200UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together