Biotinylated Human IL-10 R alpha, Fc,Avitag (ILR-H82F6) is expressed from human 293 cells (HEK293). It contains AA His 22 - Asn 235 (Accession # Q13651-1). 1 * 25 µG
Biotinylated Human IL-10 R alpha, Fc,Avitag (ILR-H82F6) is expressed from human 293 cells (HEK293). It contains AA His 22 - Asn 235 (Accession # Q13651-1). 1 * 25 µG