Biotinylated Human IL-13 R alpha 1, Fc,Avitag (IL1-H82F6) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 34 3 (Accession # NP_001551.1). 1 * 25 µG
Codice catalogo ACRBIL1-H82F6-25UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together