ContactUsLinkComponent
Mouse TNFR2, His Tag (TN2-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 23 - Gly 258 (Accession # P25119-1). 1 * 200 µG
Codice catalogo ACRBTN2-M52H8-50UG
Nome del fornitore:  ACRO Biosystems
Mouse TNFR2, His Tag (TN2-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 23 - Gly 258 (Accession # P25119-1). 1 * 200 µG
Codice catalogo ACRBTN2-M52H8-50UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together