Auftragserfassung
Switzerland
ContactUsLinkComponent
Human Alpha-fetoprotein, His Tag (ALN-H52H9) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Val 609 (Accession # P02771-1). 1 * 500 µG
Codice catalogo ACRBALN-H52H9500UG
Nome del fornitore:  ACRO Biosystems
Human Alpha-fetoprotein, His Tag (ALN-H52H9) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Val 609 (Accession # P02771-1). 1 * 500 µG
Codice catalogo ACRBALN-H52H9500UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together