Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Human IL-17E, His Tag (IL5-H52H9) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Gly 177 (Accession # NP_073626 .1). 1 * 20 µG
ACRBIL5-H52H9-20UG
:
ACRO Biosystems
Human IL-17E, His Tag (IL5-H52H9) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Gly 177 (Accession # NP_073626 .1). 1 * 20 µG
ACRBIL5-H52H9-20UG
:
ACRO Biosystems
Frequently Bought Together
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
VWR®, Divaricatori, a L, monouso
: VWR Collection
VWR®, CryoBox, cartone, 133×133 mm
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Piastra Petri, PS
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF