ContactUsLinkComponent
Human Siglec-7, Fc (L118A, G120A, E201A) Tag (SI7-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Accession # Q9Y286-1). 1 * 500 µG
  ACRBSI7-H5257500UG
Human Siglec-7, Fc (L118A, G120A, E201A) Tag (SI7-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Accession # Q9Y286-1). 1 * 500 µG
  ACRBSI7-H5257500UG
Frequently Bought Together