Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Mouse IL-17C, His Tag (NON-H5219) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # Q13253-1) . 1 * 50 µG
Catalog # ACRBNON-H5219-50UG
Supplier:
ACRO Biosystems
Expand image
Mouse IL-17C, His Tag (NON-H5219) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # Q13253-1) . 1 * 50 µG
Catalog # ACRBNON-H5219-50UG
Supplier:
ACRO Biosystems
Frequently Bought Together
Bottiglie per terreni di coltura, quadrate, sterili, Nalgene®
Supplier: Thermo Fisher Scientific
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Pipette sierologiche, Stripette™
Supplier: Corning
Fiasche per colture cellulari, Corning®
Supplier: Corning
Clear bottom black and white polystyrene microplates, 96 well, Corning®
Supplier: Corning
VWR®, CryoBox, cartone, 133×133 mm
Supplier: VWR Collection
VWR®, sigilli in pellicola di rayon traspirante per colture biologiche
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
VWR®, Cucchiai per campionamento
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Contenitori per campioni dritti con tappo a vite
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Provette per microcentrifuga Eppendorf® Protein LoBind®
Supplier: EPPENDORF