Human 4-1BB Ligand (71-254), Fc Tag, low endotoxin (active trimer) (41L-H5269) is expressed from human 293 cells (HEK293). It contai ns AA Arg 71 - Glu 254 (Accession # P41273-1). 1 * 1 mg
Codice catalogo ACRB41L-H5269-1MG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together