Accessories for PLATEMASTER®
Reload block
Codice catalogo GILSF110767
Nome del fornitore: GILSON
Vai a
Product Details & Documents
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.
Documents
Specifiche
- Descrizione:Reload block