Masterflex® Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
Katalognr. MFLX40615-73
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/4"
- Tipo di connessione:Thread to Barb
- Flow pattern:T
- Descrizione:Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
- Environmentally Preferable:
- Orifice dimensions:0,125"
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Conf:10
- Barb size/tubing int.Ø (port 3):1/4"
- Fitting material:PVDF
Frequently Bought Together