Orders LinkContactUsLinkComponent
Reactor systems, EasySyn
Reactor systems, EasySyn
EasySyn 500 advanced reactor system, 0,5 L
Codice catalogo IKAA20113434
Nome del fornitore:   IKA
Reactor systems, EasySyn
Codice catalogo IKAA20113434
Nome del fornitore:  IKA
Reactor systems, EasySyn
Reactor systems, EasySyn
EasySyn 500 advanced reactor system, 0,5 L
Price
13 890.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
EasySyn is the reactor system for organic or aqueous syntheses, and the only reactor system for which you can get all the complementary components from a single source: Overhead stirrers, circulators, vacuum pumps and controllers, sensors and automation software. All from IKA and all perfectly matched to each other. With its double-walled reactor vessels from 500 to 5000 ml, EasySyn is a medium-sized and very flexible upscaling system. It ensures safe and reproducible chemical reactions.
  • Ideal for organic or aqueous syntheses
The EasySyn reactor systems are characterised by a wide range of applications such as reflux condensations, reactions under vacuum, crystallisations, (pH-controlled) precipitation reactions, nanoparticle or catalyst syntheses as well as the transfer from laboratory to process scale.
In many applications, controllable and reproducible reaction conditions are essential to achieve reliable and comparable results. With EasySyn, these conditions are guaranteed by the use of high-quality materials in combination with technically sophisticated accessories. Our reactor systems are for example suitable for research and development, the chemical industry or materials research.
Ergonomic and safe - They are unique thanks to the ergonomically designed stand. It allows quick and easy exchange of the reactor vessels, simple alignment of the stirrer and adjustment of the working height to the user's body size.
Due to the use of borosilicate glass 3.3 and PTFE-coated stirring tools, EasySyn has a high chemical resistance and can also be used for syntheses with aggressive media such as acids or bases. The high quality of the materials used allows operation at temperatures from −50 to 200 °C and up to a vacuum of 3 mbar.
One system, many possibilities - In total, two packages are offered, each with different sizes and variations: EasySyn Starter and EasySyn Advanced. They each comprise a complete reactor system for use in the laboratory, consisting of a stand system, double-walled reactor vessel, reactor lid as well as a propeller stirrer and a suitable overhead stirrer.
The main difference between Starter and Advanced is the overhead stirrer used. EasySyn Starter includes the EUROSTAR 60 digital, the Advanced Packages the EUROSTAR 100 control, which enables additional functions, such as external control via labworldsoft® 6 (RS232 or USB interface), operation via removable remote control and programming, for example, of a timer. In addition, the Advanced systems include the LT 5.40 draining set, which enables the tempering fluid of the double jacket to be drained easily and cleanly by using special valves, thus saving valuable time when changing the reactor vessels. The Advanced System is therefore particularly suitable for applications in which the reactor vessel has to be changed or cleaned regularly.
Areas of application: Organic and aqueous syntheses; Reactions under vacuum; Crystallisations; (pH-controlled) precipitation reactions; Nanoparticle syntheses; Catalyst syntheses. Matching to this you get all the complementary components from a single source: overhead stirrers, circulators, vacuum pumps and controllers, sensors and automation software.
Specifiche
  • Descrizione:
    EasySyn 500 advanced reactor system, 0,5 L
  • Seq:
    02
  • Cat. No.:
    IKAA20113434
  • Modello:
    EasySyn 500
  • Range della temperatura di esercizio:
    5...40 °C (permissible ambient temperature)
  • Vuoto:
    3 mbar (attainable)
  • Volume utile:
    30 ml (min.)
  • L×P×A:
    500×580×950 mm
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • SY stand
    SY stand
    IKAA20103316
    Log in per disponibilità
    1 * 1 ST - SFr. 4’650.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Intervallo di temperatura
    Viscosità
    Disponibilità
    Price
  • IKAA20003521
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in per disponibilità
    1 * 10 kg - SFr. 824.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Reactor lid for the EasySyn reactor system, openings: 4×NS 29/32, 1×NS 14/23, 2×GL 18
    Reactor lid for the EasySyn reactor system, openings: 4×NS 29/32, 1×NS 14/23, 2×GL 18
    IKAA20116965
    Suitable for reactor vessels with volumes of 500 ml (SY 500 and SY 500 D) and 1000 ml (SY 1000 and SY 1000 D)
    Log in per disponibilità
    1 * 1 ST - SFr. 1’110.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Membrane vacuum pump, VACSTAR digital
    Membrane vacuum pump, VACSTAR digital
    IKAA20016236
    Log in per disponibilità
    1 * 1 ST - SFr. 3’710.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Range di temperatura
    Cost. temp.
    Disponibilità
    Price
  • IKAA4165000
    CBC 5 basic
    30...200 °C
    ±0,05 K
    Log in per disponibilità
    1 * 1 ST - SFr. 7’420.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Intervallo di temperatura
    Viscosità
    Disponibilità
    Price
  • IKAA20003520
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in per disponibilità
    1 * 10 kg - SFr. 738.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Intervallo di temperatura
    Viscosità
    Disponibilità
    Price
  • IKAA20003518
    Heating fluid
    +20...+200 °C
    50 mm²/s
    Log in per disponibilità
    1 * 9 kg - SFr. 773.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Range di temperatura
    Cost. temp.
    Disponibilità
    Price
  • IKAA4167000
    CBC 5 control
    30...200 °C
    ±0,05 K
    Log in per disponibilità
    1 * 1 ST - SFr. 8’850.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Hose set includes M 16×1 connection, PUR 90 °C, Ø 12 mm
    Hose set includes M 16×1 connection, PUR 90 °C, Ø 12 mm
    IKAA10011654
    Log in per disponibilità
    1 * 1 ST - SFr. 222.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Tube FKM, 2×1,5 m, M 16×1, −30...+180 °C, Ø 12 mm
    Tube FKM, 2×1,5 m, M 16×1, −30...+180 °C, Ø 12 mm
    IKAA4569300
    Log in per disponibilità
    1 * 1 ST - SFr. 731.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Temperature control hose, 1,6 m
    Temperature control hose, 1,6 m
    IKAA20113271
    Log in per disponibilità
    1 * 1 ST - SFr. 1’180.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Cost. temp.
    Disponibilità
    Price
  • HRC 2 temperature control
    HRC 2 temperature control
    IKAA25004524
    HRC 2 control
    0,05±K
    Log in per disponibilità
    1 * 1 ST - SFr. 7’770.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • H 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mm
    H 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mm
    IKAA20117772
    Log in per disponibilità
    1 * 1 ST - SFr. 679.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Agitatore ad asta EUROSTAR 60 digital, spina EU
    Agitatore ad asta EUROSTAR 60 digital, spina EU
    441-0326
    Log in per disponibilità
    1 * 1 ST - SFr. 2’260.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Range di temperatura
    Cost. temp.
    Disponibilità
    Price
  • 441-0408
    HBC 5 control
    20...250 °C (with integrated cooling coil*) °C
    ±0,01 K
    Log in per disponibilità
    1 * 1 ST - SFr. 6’380.00
Informazioni sul prodotto
  • Codice catalogo
    Tipo
    Range di temperatura
    Cost. temp.
    Disponibilità
    Price
  • 441-0409
    HBC 5 basic
    20...200 °C
    ±0,02 K
    Log in per disponibilità
    1 * 1 ST - SFr. 4’960.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • H 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mm
    H 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mm
    IKAA20118833
    Log in per disponibilità
    1 * 1 ST - SFr. 735.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Reactor lid for the EasySyn reactor system, openings: 3×NS 29/32, 1×NS 14/23
    Reactor lid for the EasySyn reactor system, openings: 3×NS 29/32, 1×NS 14/23
    IKAA20113649
    Suitable for reactor vessels with volumes of 500 ml (SY 500 and SY 500 D) and 1000 ml (SY 1000 and SY 1000 D)
    Log in per disponibilità
    1 * 1 ST - SFr. 666.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    IKAA20017366
    All configurations with any number of devices can be controlled at the same time
    Log in per disponibilità
    1 * 1 ST - SFr. 15’210.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Valvola di scarico inferiore con sensore di temperatura PT 100 integrato
    Valvola di scarico inferiore con sensore di temperatura PT 100 integrato
    IKAA20121894
    Log in per disponibilità
    1 * 1 ST - SFr. 1’190.00