Inserts for U-D master manual samplers
U-D Master insert split, Acciaio inossidabile 316L, 1,5 ml
Codice catalogo SAMP1230T-15S
Nome del fornitore: Sampling Systems
Inserts for U-D master manual samplers
Codice catalogo SAMP1230T-15S
Nome del fornitore: Sampling Systems
Vai a
Product Details & Documents
Stainless steel inserts for U-D Master manual samplers.
- Made from 316 stainless steel
These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.
Specifiche
- Volume:1,5 ml
- Descrizione:U-D Master insert split
- Materiale:Acciaio inossidabile 316L
- Seq:04
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.