ContactUsLinkComponent
PERMABOND17X35MM-TARE CLEAR GLASS 16X35MM STRAIGHT WALL HIGH RECOVERY VIALS WITH 18-400 GPI THREAD FINISH. COMES WITH PERMABOND 17X3 5MM CERAMIC BARCODE/TEXT LABELS TO CUSTOMER SPECIFICATION ON SIDE OF VIAL. VIALS WILL BE TARE WEIGHED TO 4 DECIMAL PL 1 * 1 ST
Catalog # TRADSWHR1635C
Supplier:  TRADEWINDS DIRECT
PERMABOND17X35MM-TARE CLEAR GLASS 16X35MM STRAIGHT WALL HIGH RECOVERY VIALS WITH 18-400 GPI THREAD FINISH. COMES WITH PERMABOND 17X3 5MM CERAMIC BARCODE/TEXT LABELS TO CUSTOMER SPECIFICATION ON SIDE OF VIAL. VIALS WILL BE TARE WEIGHED TO 4 DECIMAL PL 1 * 1 ST
Catalog # TRADSWHR1635C
Supplier:  TRADEWINDS DIRECT
Frequently Bought Together