Orders LinkContactUsLinkComponent
FUNNEL, WEIGHING, BOROSILICATE, 6ML SOLID CAPACITY, 25ML LIQUID CAPACIT
FUNNEL, WEIGHING, BOROSILICATE, 6ML SOLID CAPACITY, 25ML LIQUID CAPACIT
Codice catalogo CGLACG-1760-30
Marketsource
Nome del fornitore:   CHEMGLASS
FUNNEL, WEIGHING, BOROSILICATE, 6ML SOLID CAPACITY, 25ML LIQUID CAPACIT
Codice catalogo CGLACG-1760-30
Nome del fornitore:  CHEMGLASS
Marketsource
FUNNEL, WEIGHING, BOROSILICATE, 6ML SOLID CAPACITY, 25ML LIQUID CAPACIT
Price
SFr. 83.00
Log in per disponibilità
1 * 6 ST