Masterflex® Quick-Disconnect Body, Acetal, 1/8" Flow, Valved, 1/4" Hose Barb
Codice catalogo MFLX70000-27
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Disconnect and reconnect your flow path with ease.
- Audible click lets you know the coupling is secure
- Many hose barb sizes to choose from
- Valved design allows you to cut off fluid flow when the fittings are disconnected
- Material of construction is acetal│Has a 1/4" hose barb
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and the ability to be gamma sterilized.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):0,25"
- Tipo di connessione:Body
- Descrizione:Masterflex Quick-Disconnect Body, Acetal, ⅛" Flow, Valved, ¼" Hose Barb
- Seq:04
- Environmentally Preferable:
- Modello:70000-27
- Conf:1
- Fitting material:Acetal
- Quick-disconnect flow size:0,13"
Frequently Bought Together