Fitting, Black Nylon, Elbow, Hose Barb Union, 1/16"; 10/Pk
Codice catalogo MFLX41518-46
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Dimensioni del raccordo/Ø int. del tubo (porta 1):1/16"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/16"
- Tipo di connessione:Barbed Union
- Flow pattern:Straight
- Descrizione:Fitting, Black Nylon, Elbow, Hose Barb Union, 1/16"; 10/Pk
- Seq:18
- Environmentally Preferable:
- Modello:41518-46
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:Nylon
Frequently Bought Together