Sanitary Clamp Adapter, 1/4" NPT(M) X 1/4" Clamp
Codice catalogo MFLX34031-04
Nome del fornitore: Avantor Fluid Handling
CE certificate
Informazioni su questo articolo
Create a secure connection in your sanitary fitting system.
- Constructed of chemically inert PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
- Tipo di connessione:Sanitary to Thread
- Flow pattern:Straight
- Sanitary clamp size (Port 1):1/4"
- Thread size NPT(M) (port 2):1/4"
- Descrizione:Sanitary Clamp Adapter, 1/4" NPT(M)×1/4" Clamp
- Environmentally Preferable:
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Garanzia:2 years
- Conf:1
- Fitting material:PTFE
- Sanitary gasket size:1/4"
Frequently Bought Together