Masterflex® Barbed to Sanitary Adapter, ¹/₂" ID to 1¹/₂", ADCF PP; 10/PK, Avantor®
Codice catalogo MFLX31515-92
Nome del fornitore: Avantor Fluid Handling
CE certificate
Informazioni su questo articolo
Certificate of compliance included!
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Animal derivative component free
- Meet USP Class IV requirements
- Sterilisable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow - and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilised by gamma irradiation. Material meets the requirements for USP Class VI.
Delivery: Includes certificate of compliance.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/2"
- Tipo di connessione:Sanitary to Barb
- Flow pattern:Straight
- Sanitary clamp size (Port 1):1-1/2"
- Descrizione:Barbed to Sanitary Adapter, ¹/₂" ID to 1-1/2", ADCF PP; 10/PK, Avantor®
- Seq:06
- Environmentally Preferable:
- Sterilizzazione:Ethylene oxide (ETO) or gamma
- Conf:10
- Fitting material:Polypropylene
- Sanitary gasket size:1-1/2"
Sanitary Caps