Masterflex® Gold-colored IDentifier Hinge Clamp, 2" Clamp Size, 304 SS; 1/EA
Codice catalogo MFLX30562-53
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Easy visual identification of critical joints for review in your process.
- Constructed of gold-coloured 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-coloured sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.
- Tipo di collegamento:Sanitary
- Descrizione:Gold-coloured Identifier Hinge Clamp, 2" Clamp Size, 304 SS; 1/EA
- Materiale:Acciaio inossidabile 304
- Environmentally Preferable:
- Sanitary clamp size (port 1):2"
- Garanzia:1 year
- Conf:1
- Tipo di morsetto:Sanitary
Frequently Bought Together