Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, L/S® 13
Codice catalogo MFLX07013-91
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07013-10 and -21
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Descrizione:Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, L/S® 13 Tubing
- Seq:04
- Environmentally Preferable:
- Modello:07013-91
- Open-head sensor:No
- Tubing sizes accepted:L/S 13
- Garanzia:2 years
- Conf:1
- Housing material:PC
- Rotor material:Stainless Steel
Frequently Bought Together