Masterflex® L/S® Standard Digital Pump Systems, Avantor®
L/S Digital Pump System with Easy-Load 3 Pump Head, 600 RPM, 115/230V, CH, 480 ml/min, ABS plastic and aluminium, 77800-60 (L/S® Easy-Load 3 Pump Head), 215,9 mm, 352,6 mm, 210,8 mm, 600 rpm, 90 - 260 VAC; 50/60 Hz; 2,2 A @ 115 V; 1,1 A @ 230 V
Codice catalogo MFLX77921-65-CH
2 year warranty
CE Compliant
Nome del fornitore: Avantor Fluid Handling
Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Codice catalogo MFLX77921-65-CH
Nome del fornitore: Avantor Fluid Handling
2 year warranty
CE Compliant
Vai a
Product Details & Documents
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
- Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
- Select your preferred language from the user-friendly menu for programming and operation
- Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures dispensing accuracy
- Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
- Models with open-head sensor stop drive motor when pump head is opened
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Specifiche
- Massima velocità del flusso:480 ml/min
- Potenza:90 - 260 VAC; 50/60 Hz; 2,2 A @ 115 V; 1,1 A @ 230 V
- Descrizione:L/S Digital Pump System with Easy-Load 3 Pump Head, 600 RPM, 115/230V
- Altezza:215,9 mm
- Lunghezza:352,6 mm
- Seq:24
- Testa pompa:77800-60 (L/S® Easy-Load 3 Pump Head)
- Larghezza:210,8 mm
- Housing material:ABS plastic and aluminium
- Electrical plug type:CH
- Max. RPM:600 rpm
- Tensione:90 - 260 VAC
- Min. flow rate per channel:0,08 ml/min
- Open-head sensor:No
- Connesso cloud:No
- Remote I/O:4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- Tubo:06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- Digital dispensing:Yes
- Garanzia:2 years
- Drive IP rating:IP 33
- Modello:77921-65
- Washdown capable:No
- Min. RPM:0,1 rpm
- Numero di rulli:3
- Ciclo di funzionamento:Continuous
- Corrente:2,2 A @ 115 V; 1,1 A @ 230 V
- Max. flow rate per channel:480 ml/min
- Frequenza:50/60 Hz
- Reversible motor:Yes
- Portata di flusso min.:0,08 ml/min
- Gruppo motore:07522-20 (L/S® Standard Digital Drive, 600 rpm)
- Cat. No.:MFLX77921-65-CH
- Antigoccia:Yes
- N. di canali:1
- Pump series:L/S
- Accuratezza:±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
- Tubing sizes accepted:L/S® 13, 14, 16, 25, 17, 18
- Tipo di controllo:Variable-Speed Digital
Services
Informazioni sul prodotto
- Codice catalogoDescrizioneDisponibilitàPrice
- Qualifica dell'installazione e qualifica del funzionamento della pompa peristaltica presso la sede del cliente secondo i protocolli SOP (Standard Operating Procedure) Avantor. Protocolli di qualifica SOP Avantor inclusi. Spese di viaggio incluse (escluso l'eventuale costo del viaggio aereo).Log in per disponibilità1 * 1 Un. - SFr. 1’670.00