[EN]CONCEPT FRAME HEIGHT ADJUSTMENT WITH 1 * 1 pezzi
Catalog # SOVE10149032
Supplier: TRESTON
Frequently Bought Together
[EN]CUBIS II HC MCA66P-3CEU 1 MDS 1 * 1 pezzi
Supplier: Sartorius Balances
[EN]CUBIS II HC MCA116S-3S00 2 3 ION 1 * 1 pezzi
Supplier: Sartorius Balances
VWR®, Blocchi riscaldanti modulari e accessori per sistema di riscaldamento a blocchi
Supplier: VWR Collection
High temperature tube furnaces with Tmax 1600 °C, RHTC series
Supplier: NABERTHERM
[EN]MICROTITRE PLATE BLOCK INSERT 1 * 1 pezzi
Supplier: Cole-Parmer
Pesi di calibrazione, OIML M1
Supplier: OHAUS
VWR®, Accessories for VWR Standard and Advanced Shakers
Supplier: VWR Collection
Cubis® II MCA 226S Ultra-High Resolution Semi-Micro Balances
Supplier: Sartorius Balances
[EN]SHELF PERFORATED STAINLESS STEEL FOR 1 * 1 pezzi
Supplier: MEMMERT
[EN]CUBIS II HC MCA66S-3CEU QP99 ION 1 * 1 pezzi
Supplier: Sartorius Balances
[EN]CUBIS II HC MCA66P-3CEU 2 3 4 ION 1 * 1 pezzi
Supplier: Sartorius Balances
VWR® TA Professional, Analytical Balances with Touch Screen
Supplier: VWR Collection