Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent

Cell Lysis Solution, Ultrapure Bioreagent, Plastic bottle, 10 mL
Codice catalogo X193-01
   Più sicuro dal punto di vista della salute umana e dell'ambiente
Nome del fornitore:   Avantor
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Codice catalogo X193-01
Nome del fornitore:  Avantor
   Più sicuro dal punto di vista della salute umana e dell'ambiente
Numero CAS:  
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent

Cell Lysis Solution, Ultrapure Bioreagent, Plastic bottle, 10 mL

Numero CAS:  
Price
SFr. 173.00
Log in per disponibilità
1 * 10 mL
Product Details & Documents

J.T. Baker® Cell Lysis Solution. Optimized multi-component, ready-to-use solution for extraction and recovery of AAV particles from HEK293 or SF9 cells. The product offers quantitative recovery through efficient cell lysis and protection of vector particles against shear stress. The solution is formulated with non-ionic detergents which are octyl phenol ethoxylate free.

  • Comparable cell lysis performance to octyl phenol ethoxylate
  • Improved protection of viral particles
  • Reduced foaming and viscosity
  • 0.2 µm filtered solution

Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)

Chemical Specifications of Non-GMP Cell lysis solution:
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total microbial count: No growth detected
UV-Vis (max Absorption) at 380-450 nm: For information only

Specifiche

Specifiche

  • Tipo di conf.:
    Plastic bottle
  • Conf:
    10 mL
  • Descrizione:
    Cell Lysis Solution, Ultrapure Bioreagent
  • Cat. No.:
    X193-01
  • Environmentally Preferable:
    Più sicuro dal punto di vista della salute umana e dell'ambiente
  • Certificazione ambientale:
    EPA safer choice chemical

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.