Masterflex® luer fitting, male luer lock x 1/4 hose barb, Violet, 25/pk"
Catalog # MFLX45520-46
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):1/4"
- Tipo di connessione:Male Luer to Barb
- Flow pattern:Straight
- Descrizione:luer fitting, male luer lock×1/4 hose barb, Violet, 25/pk"
- Environmentally Preferable:
- Connection type (port 1):Male luer lock
- No. of ports:2
- Conf:25
- Fitting material:CrystalVu
Frequently Bought Together