Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
luer fitting, male luer lock×1/16 hose barb, Berry, 25/pk", 1/16", Male Luer to Barb, CrystalVu, Straight
Codice catalogo MFLX45520-01
Nome del fornitore: Avantor Fluid Handling
Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Codice catalogo MFLX45520-01
Nome del fornitore: Avantor Fluid Handling
Vai a
Product Details & Documents
Quick and easy assembly without tools.
- Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
Specifiche
- Dimensioni del raccordo/Ø int. del tubo (porta 1):1/16"
- Tipo di connessione:Male Luer to Barb
- Flow pattern:Straight
- Descrizione:luer fitting, male luer lock×1/16 hose barb, Berry, 25/pk"
- Fitting material:CrystalVu
- Cat. No.:MFLX45520-01
- Connection type (port 1):Male luer lock
- No. of ports:2