Cadence Luer Fitting, Nickel-Plated Brass, Male Luer Lock to Male Luer Lock
Codice catalogo MFLX41507-55
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Excellent durability.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
- Full material traceability on all items
Constructed of durable plated brass, these Luer Adapter Fittings are ideal for a wide range of applications.
- Tipo di connessione:Male Luer Union
- Flow pattern:Straight
- Descrizione:Luer Fitting, Nickel-Plated Brass, Male Luer Lock to Male Luer Lock
- Environmentally Preferable:
- Connection type (port 1):Male luer lock
- Connection type (port 2):Male luer lock
- No. of ports:2
- Conf:1
- Fitting material:Nickel-Plated Brass
Frequently Bought Together