Threaded Reducing Bushing, 1" NPT(M) x 3/4" NPT(F), PTFE
Catalog # MFLX34030-54
Supplier: Avantor Fluid Handling
About this item
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.
- Tipo di connessione:Threaded Reducer
- Flow pattern:Straight
- Thread size NPT(F) (port 2):¾"
- Thread size NPT(M) (port 1):1"
- Descrizione:Threaded Reducing Bushing, 1" NPT(M) × ¾" NPT(F), PTFE
- Seq:50
- Environmentally Preferable:
- Modello:34030-54
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Conf:1
- Fitting material:PTFE
Frequently Bought Together