Masterflex® Fitting, Brass, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/4" NPT(M); 5/PK
Codice catalogo MFLX30904-10
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):¼"
- Tipo di connessione:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¼"
- Descrizione:Fitting, Brass, Straight, Hosebarb to Thread Adapter, ¼" ID × ¼" NPT(M); 5/PK
- Seq:05
- Environmentally Preferable:
- Modello:30904-10
- No. of ports:2
- Garanzia:1 year
- Conf:5
- Fitting material:Brass
Frequently Bought Together