Masterflex® Fitting, Polypropylene, Elbow, Compression to Threaded, 3/8" OD x 3/8" NPT(M); 10/PK
Codice catalogo MFLX06386-50
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Maintain a permanent connection with these adapters.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
- Tipo di connessione:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):⅜"
- Descrizione:Compression male pipe adapter elbow, ⅜" × ⅜", polypropylene, 10 Pack
- Seq:12
- Environmentally Preferable:
- Modello:06386-50
- Orifice dimensions:8,73 mm (0,34375")
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:Polypropylene
Frequently Bought Together