CPC® Miniature Quick-Disconnect Fitting, Hose Barb Complete Coupling, Acetal, Straight-Through, 1/16" ID, SMC01; 10/PK
Catalog # MFLX06363-88
Supplier: Avantor Fluid Handling
About this item
No more kinked tubing - allows tubing to rotate freely.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
- Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle ¹/₁₆" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
The complete couplings come with one insert and one body fitting.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):¹⁄₁₆"
- Tipo di connessione:Quick-Disconnect to Barb
- Flow pattern:Straight
- Temperatura massima:82 °C (180 °F)
- Descrizione:Miniature Quick-Disconnect Fitting, Hose Barb Complete Coupling, Acetal, Straight-Through, ¹/₁₆" ID, SMC01; 10/PK
- Environmentally Preferable:
- O-Ring material:Buna N
- Valve material:None
- Temperatura minima:–40 °C (–40 °F)
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Garanzia:1 year
- Conf:10
- Fitting material:Acetal
- Quick-disconnect flow size:³⁄₃₂"
Frequently Bought Together