Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/8" ID x 1/16" ID; 25/PK
Codice catalogo MFLX50622-23
Nome del fornitore: Avantor Fluid Handling
CE certificate
Informazioni su questo articolo
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):⅛"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):¹⁄₁₆"
- Tipo di connessione:Barbed Reducer
- Flow pattern:Straight
- Descrizione:Fitting, polypropylene, straight, hose barb reducer, int.Ø ⅛×¹⁄₁₆";10/pack
- Seq:48
- Environmentally Preferable:
- Modello:50622-23
- Sterilizzazione:Ethylene Oxide (ETO) or Gamma
- Conf:25
- Fitting material:Polypropylene
Frequently Bought Together