Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 1/4-28 UNF; 25/PK
Catalog # MFLX45518-82
Supplier: Avantor Fluid Handling
CE certificate
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Connection Type: Male Luer Lock to Threaded, 1/4-28 UNF
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Tipo di connessione:Male Luer to Thread
- Flow pattern:Straight
- Descrizione:Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, ¼-28 UNF; 25/PK
- Seq:07
- Environmentally Preferable:
- Modello:45518-82
- Connection type (port 1):Male luer lock
- Connection type (port 2):¼-28 UNF
- No. of ports:2
- Conf:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 1):¼"-28
Frequently Bought Together