Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 3/32" ID x 10-32 UNF(M); 10/PK
Catalog # MFLX40629-00
Supplier: Avantor Fluid Handling
CE certificate
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/32" ID x 10-32 UNF(M)
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Dimensioni del raccordo/Ø int. del tubo (porta 2):3/32"
- Tipo di connessione:Thread to Barb
- Flow pattern:Straight
- Descrizione:Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 3/32" ID x 10-32 UNF(M); 10/PK
- Seq:19
- Environmentally Preferable:
- Modello:40629-00
- Sterilizzazione:Ethylene Oxide (ETO)
- Conf:10
- Fitting material:HDPE
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together