Masterflex® Fitting, Nylon, Elbow, Hose Barb Union, 3/32"; 10/PK
Catalog # MFLX40623-16
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Dimensioni del raccordo/Ø int. del tubo (porta 1):3/32"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):3/32"
- Tipo di connessione:Barbed Union
- Flow pattern:Elbow
- Descrizione:Fitting, Nylon, Elbow, Hose Barb Union, 3/32"; 10/PK
- Seq:15
- Environmentally Preferable:
- Modello:40623-16
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:Nylon
Frequently Bought Together