Masterflex® Fitting, Polypropylene, Tee, Hose Barb Union, 1/8"ID; 10/PK
Codice catalogo MFLX40613-47
Nome del fornitore: Avantor Fluid Handling
CE certificate
Informazioni su questo articolo
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/8"
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Dimensioni del raccordo/Ø int. del tubo (porta 1):⅛"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):⅛"
- Tipo di connessione:Barbed Union
- Flow pattern:T
- Descrizione:Fitting, Polypropylene, Tee, Hose Barb Union, ⅛"ID; 10/PK
- Seq:28
- Environmentally Preferable:
- Modello:40613-47
- Sterilizzazione:Ethylene Oxide (ETO) or Autoclave
- Conf:10
- Barb size/tubing int.Ø (port 3):⅛"
- Fitting material:Polypropylene
Frequently Bought Together