Masterflex® Fitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 25/PK
Catalog # MFLX30800-00
Supplier: Avantor Fluid Handling
CE certificate
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
- Tipo di connessione:Female Luer to Thread
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/8"
- Descrizione:Fitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 25/PK
- Seq:01
- Environmentally Preferable:
- Modello:30800-00
- Connection type (port 1):Female luer
- Connection type (port 2):1/8-27-NPT thread
- No. of ports:2
- Garanzia:3 Months
- Conf:25
- Fitting material:Polypropylene
Frequently Bought Together