Masterflex® Fitting, Nylon, Straight, Hose Barb Reducer, 3/8" ID x 1/4" ID; 100/PK
Codice catalogo MFLX30704-89
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range −50 to +250 °F (−46 to +121 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):3/8"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/4"
- Tipo di connessione:Barbed Reducer
- Flow pattern:Straight
- Descrizione:Fitting, Nylon, Straight, Hose Barb Reducer, 3/8" ID x 1/4" ID; 100/PK
- Seq:34
- Environmentally Preferable:
- Modello:30704-89
- Sterilizzazione:Ethylene Oxide (ETO)
- Conf:100
- Fitting material:Nylon
Frequently Bought Together