Masterflex® Fitting, HDPE, Straight, Hose Barb Union, 1/4" ID; 100/PK
Codice catalogo MFLX30625-85
Nome del fornitore: Avantor Fluid Handling
CE certificate
Informazioni su questo articolo
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
- Connection Type: 1/4" ID
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):¼"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):¼"
- Tipo di connessione:Barbed Union
- Flow pattern:Straight
- Descrizione:Fitting, HDPE, Straight, Hose Barb Union, ¼" ID; 100/PK
- Seq:17
- Environmentally Preferable:
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:100
- Fitting material:HDPE
Frequently Bought Together